Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036898.2 | NP_001031975.1 | 6 - 7 | 606 | 71 | |
NM_123047.4 | NP_198505.2 | 6 - 7 | 607 | 71 | ||
GenBank mRNA | AK175760.1 | - | 6 - 7 | 607 | 71 | |
BT010611.1 | - | 6 - 7 | 467 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |