Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123064.3 | NP_568545.1 | 1 - 2 | 227 | 154 | |
GenBank mRNA | AF378857.1 | - | 1 - 2 | 154 | 154 | |
AK175306.1 | - | 1 - 2 | 144 | 154 | ||
AY050464.1 | - | 1 - 2 | 101 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |