Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344166.1 | NP_001318683.1 | 3 - 4 | 649 | 69 | |
NM_123077.3 | NP_198534.3 | 1 - 2 | 229 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |