Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344168.1 | NP_001331331.1 | 5 - 6 | 1255 | 77 | |
NM_001344169.1 | NP_001331332.1 | 5 - 6 | 1212 | 77 | ||
NM_123078.3 | NP_568546.1 | 5 - 6 | 1255 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |