Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344178.1 | NP_001331811.1 | 2 - 3 | 857 | 129 | |
NM_001344179.1 | NP_001331810.1 | 2 - 3 | 857 | 129 | ||
NM_001344180.1 | NP_001331809.1 | 2 - 3 | 857 | 129 | ||
NM_123089.3 | NP_568547.1 | 2 - 3 | 787 | 129 | ||
GenBank mRNA | AY056316.1 | - | 2 - 3 | 778 | 129 | |
BT015776.1 | - | 2 - 3 | 769 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |