Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSR 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123119.4 | NP_198576.1 | 7 - 8 | 984 | 92 | |
GenBank mRNA | AF419608.1 | - | 7 - 8 | 828 | 92 | |
AF428386.1 | - | 7 - 8 | 828 | 92 | ||
AY079113.1 | - | 7 - 8 | 755 | 92 | ||
BT000753.1 | - | 7 - 8 | 835 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |