Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085206.1 | NP_001078675.1 | 1 - 2 | 778 | 105 | |
NM_123125.4 | NP_198582.2 | 1 - 2 | 961 | 105 | ||
GenBank mRNA | AK229025.1 | - | 1 - 2 | 778 | 105 | |
BT022067.1 | - | 1 - 2 | 749 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |