Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123142.5 | NP_198599.1 | 2 - 3 | 3678 | 90 | |
GenBank mRNA | AY102096.1 | - | 2 - 3 | 3589 | 90 | |
BT004510.1 | - | 1 - 2 | 3486 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |