Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085208.2 | NP_001078677.1 | 10 - 11 | 916 | 110 | |
NM_001203503.1 | NP_001190432.1 | 9 - 10 | 725 | 110 | ||
NM_001344222.1 | NP_001331291.1 | 9 - 10 | 1008 | 110 | ||
NM_123144.4 | NP_198601.2 | 10 - 11 | 883 | 110 | ||
GenBank mRNA | AF404865.2 | - | 9 - 10 | 687 | 110 | |
AY084419.1 | - | 9 - 10 | 717 | 110 | ||
AY136333.1 | - | 10 - 11 | 775 | 110 | ||
BT008815.1 | - | 9 - 10 | 687 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |