Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111257.4 | NP_566214.1 | 2 - 3 | 746 | 112 | |
GenBank mRNA | AY059721.1 | - | 2 - 3 | 622 | 112 | |
AY091351.1 | - | 2 - 3 | 355 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |