Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111280.5 | NP_187059.1 | 1 - 2 | 506 | 89 | |
GenBank mRNA | AK226432.1 | - | 1 - 2 | 360 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |