Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111283.4 | NP_187062.1 | 7 - 8 | 698 | 80 | |
GenBank mRNA | AK226804.1 | - | 695 | 80 | ||
AY052267.1 | - | 7 - 8 | 638 | 80 | ||
AY060521.1 | - | 6 - 7 | 563 | 80 | ||
AY140084.1 | - | 7 - 8 | 641 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |