Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337509.1 | NP_001326045.1 | 12 - 13 | 1476 | 89 | |
NM_111295.4 | NP_187074.1 | 18 - 19 | 2076 | 89 | ||
GenBank mRNA | AF441079.1 | - | 18 - 19 | 1994 | 89 | |
AK226884.1 | - | 18 - 19 | 2019 | 89 | ||
AY090938.1 | - | 18 - 19 | 2015 | 89 | ||
AY117340.1 | - | 18 - 19 | 1865 | 89 | ||
KJ138841.1 | - | 18 - 19 | 1865 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |