Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337509.1 | NP_001326045.1 | 15 - 16 | 1848 | 71 | |
NM_111295.4 | NP_187074.1 | 21 - 22 | 2448 | 71 | ||
GenBank mRNA | AF441079.1 | - | 21 - 22 | 2366 | 71 | |
AK226884.1 | - | 21 - 22 | 2391 | 71 | ||
AY090938.1 | - | 21 - 22 | 2387 | 71 | ||
AY117340.1 | - | 21 - 22 | 2237 | 71 | ||
KJ138841.1 | - | 21 - 22 | 2237 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |