Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111297.4 | NP_187076.2 | 8 - 9 | 1369 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |