Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339086.1 | NP_001327912.1 | 2 - 3 | 300 | 82 | |
NM_001339087.1 | NP_001327910.1 | 2 - 3 | 191 | 82 | ||
NM_001339088.1 | NP_001327911.1 | 1 - 2 | 53 | 82 | ||
NM_114134.2 | NP_189853.1 | 1 - 2 | 53 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |