Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114147.5 | NP_189865.1 | 3 - 4 | 534 | 102 | |
GenBank mRNA | AY062851.1 | - | 3 - 4 | 499 | 102 | |
AY114574.1 | - | 3 - 4 | 360 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |