Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339091.1 | NP_001326640.1 | 9 - 10 | 1772 | 126 | |
NM_114187.5 | NP_566865.2 | 10 - 11 | 1954 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |