Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339092.1 | NP_001319676.1 | 9 - 10 | 1029 | 60 | |
NM_001339093.1 | NP_001327900.1 | 9 - 10 | 1203 | 60 | ||
NM_001339094.1 | NP_001327899.1 | 8 - 9 | 1210 | 60 | ||
NM_001339095.1 | NP_001327901.1 | 7 - 8 | 1255 | 60 | ||
NM_114189.3 | NP_189907.2 | 8 - 9 | 1048 | 60 | ||
GenBank mRNA | AB088121.1 | - | 7 - 8 | 760 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |