Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203077.1 | NP_001190006.1 | 24 - 25 | 3678 | 85 | |
NM_001339097.1 | NP_001327198.1 | 24 - 25 | 3862 | 85 | ||
NM_114198.4 | NP_189916.4 | 24 - 25 | 3672 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |