Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JAZ11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114212.5 | NP_189930.1 | 4 - 5 | 478 | 84 | |
GenBank mRNA | BT015154.1 | - | 4 - 5 | 368 | 84 | |
BT015661.1 | - | 4 - 5 | 354 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Arabidopsis thaliana Probe spans exons |