Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161190.2 | NP_001154662.2 | 16 - 17 | 2788 | 70 | |
NM_001161191.3 | NP_001154663.2 | 16 - 17 | 2906 | 70 | ||
NM_001339114.1 | NP_001325812.1 | 16 - 17 | 2916 | 70 | ||
NM_001339115.1 | NP_001325811.1 | 16 - 17 | 2919 | 70 | ||
NM_001339116.1 | NP_001325813.1 | 11 - 12 | 2289 | 70 | ||
NM_114260.1 | NP_189978.1 | 14 - 15 | 2641 | 70 | ||
GenBank mRNA | DQ479972.1 | - | 14 - 15 | 2641 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |