Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161190.2 | NP_001154662.2 | 18 - 19 | 3221 | 65 | |
NM_001161191.3 | NP_001154663.2 | 18 - 19 | 3339 | 65 | ||
NM_001339114.1 | NP_001325812.1 | 18 - 19 | 3349 | 65 | ||
NM_001339115.1 | NP_001325811.1 | 18 - 19 | 3352 | 65 | ||
NM_001339116.1 | NP_001325813.1 | 13 - 14 | 2722 | 65 | ||
NM_114260.1 | NP_189978.1 | 16 - 17 | 3074 | 65 | ||
GenBank mRNA | DQ479972.1 | - | 16 - 17 | 3074 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |