Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161190.2 | NP_001154662.2 | 10 - 11 | 1213 | 124 | |
NM_001161191.3 | NP_001154663.2 | 10 - 11 | 1331 | 124 | ||
NM_001339114.1 | NP_001325812.1 | 10 - 11 | 1341 | 124 | ||
NM_001339115.1 | NP_001325811.1 | 10 - 11 | 1344 | 124 | ||
NM_001339116.1 | NP_001325813.1 | 5 - 6 | 714 | 124 | ||
NM_114260.1 | NP_189978.1 | 8 - 9 | 1066 | 124 | ||
GenBank mRNA | DQ479972.1 | - | 8 - 9 | 1066 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |