Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035731.2 | NP_001030808.1 | 22 - 23 | 3090 | 69 | |
NM_001339129.1 | NP_001326943.1 | 21 - 22 | 3209 | 69 | ||
NM_001339130.1 | NP_001326945.1 | 22 - 23 | 3110 | 69 | ||
NM_001339131.1 | NP_001326944.1 | 22 - 23 | 3093 | 69 | ||
NM_114302.3 | NP_566869.1 | 22 - 23 | 3080 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |