Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035731.2 | NP_001030808.1 | 4 - 5 | 1298 | 106 | |
NM_001339129.1 | NP_001326943.1 | 3 - 4 | 1417 | 106 | ||
NM_001339130.1 | NP_001326945.1 | 4 - 5 | 1318 | 106 | ||
NM_001339131.1 | NP_001326944.1 | 4 - 5 | 1301 | 106 | ||
NM_114302.3 | NP_566869.1 | 4 - 5 | 1288 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |