Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339152.1 | NP_001319682.1 | 4 - 5 | 542 | 152 | |
NM_001339153.1 | NP_001327210.1 | 4 - 5 | 1137 | 152 | ||
NM_114322.5 | NP_190040.3 | 4 - 5 | 1137 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |