Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203083.1 | NP_001190012.1 | 6 - 7 | 648 | 76 | |
NM_114344.4 | NP_566872.1 | 6 - 7 | 980 | 76 | ||
GenBank mRNA | AF195545.1 | - | 6 - 7 | 639 | 76 | |
AY065396.1 | - | 6 - 7 | 653 | 76 | ||
AY096729.1 | - | 5 - 6 | 591 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |