Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114362.3 | NP_190079.2 | 2 - 3 | 1389 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |