Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339194.1 | NP_001327790.1 | 5 - 6 | 822 | 76 | |
NM_001339195.1 | NP_001327789.1 | 4 - 5 | 729 | 76 | ||
NM_001339196.1 | NP_001327788.1 | 4 - 5 | 929 | 76 | ||
NM_001339197.1 | NP_001327787.1 | 5 - 6 | 822 | 76 | ||
NM_114382.5 | NP_190099.3 | 4 - 5 | 710 | 76 | ||
GenBank mRNA | AB247155.1 | - | 4 - 5 | 670 | 76 | |
DQ508794.2 | - | 4 - 5 | 670 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |