Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SYP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339202.1 | NP_001319688.1 | 7 - 8 | 748 | 95 | |
NM_114397.2 | NP_566878.1 | 7 - 8 | 929 | 95 | ||
GenBank mRNA | AF355758.1 | - | 7 - 8 | 748 | 95 | |
DQ446736.1 | - | 7 - 8 | 712 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |