Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118506.3 | NP_194106.1 | 1 | 1637 | 141 | |
NM_202870.1 | NP_974599.1 | 2 | 1354 | 141 | ||
GenBank mRNA | AY057475.1 | - | 1 | 1565 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |