Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 17 - 18 | 2289 | 98 | |
NM_179601.1 | NP_849932.1 | 17 - 18 | 2134 | 98 | ||
GenBank mRNA | AF436826.1 | - | 17 - 18 | 2143 | 98 | |
AX765864.1 | - | 17 - 18 | 2060 | 98 | ||
AY053403.1 | - | 17 - 18 | 2128 | 98 | ||
BT002234.1 | - | 17 - 18 | 2060 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |