Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126440.3 | NP_178488.1 | 2 - 3 | 906 | 62 | |
GenBank mRNA | BT015108.1 | - | 2 - 3 | 794 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |