Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126478.4 | NP_178526.1 | 8 - 9 | 943 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |