Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126484.5 | NP_178532.2 | 7 - 8 | 1086 | 102 | |
GenBank mRNA | AY099564.1 | - | 7 - 8 | 990 | 102 | |
BT001247.1 | - | 7 - 8 | 972 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |