Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126486.6 | NP_178534.2 | 3 - 4 | 754 | 63 | |
NM_201689.3 | NP_973418.2 | 3 - 4 | 587 | 63 | ||
GenBank mRNA | AY337455.1 | - | 3 - 4 | 540 | 63 | |
BT004283.1 | - | 3 - 4 | 543 | 63 | ||
BT005609.1 | - | 3 - 4 | 409 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |