Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AtPOT1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124798.2 | NP_001118270.1 | 10 - 11 | 1778 | 82 | |
NM_001124799.2 | NP_001118271.1 | 10 - 11 | 1589 | 82 | ||
NM_126547.5 | NP_178592.3 | 10 - 11 | 1686 | 82 | ||
GenBank mRNA | AK221471.1 | - | 10 - 11 | 1455 | 82 | |
AY884593.2 | - | 8 - 9 | 1037 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |