Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161025.1 | NP_001154497.1 | 4 - 5 | 333 | 66 | |
NM_126586.4 | NP_178631.1 | 2 - 3 | 224 | 66 | ||
GenBank mRNA | AB073178.1 | - | 2 - 3 | 161 | 66 | |
AK227339.1 | - | 2 - 3 | 201 | 66 | ||
BT003090.1 | - | 2 - 3 | 94 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |