Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335289.1 | NP_001318204.1 | 9 - 10 | 936 | 124 | |
NM_001335290.1 | NP_001324184.1 | 8 - 9 | 756 | 124 | ||
NM_001335291.1 | NP_001324181.1 | 9 - 10 | 805 | 124 | ||
NM_001335292.1 | NP_001324182.1 | 9 - 10 | 801 | 124 | ||
NM_001335293.1 | NP_001324183.1 | 9 - 10 | 788 | 124 | ||
NM_126614.3 | NP_565332.1 | 9 - 10 | 810 | 124 | ||
GenBank mRNA | AY086482.1 | - | 9 - 10 | 710 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |