Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126675.3 | NP_565338.1 | 4 - 5 | 1704 | 70 | |
GenBank mRNA | BT005794.1 | - | 4 - 5 | 1473 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |