Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335311.1 | NP_001325220.1 | 3 - 4 | 945 | 76 | |
NM_126681.3 | NP_178722.1 | 3 - 4 | 747 | 76 | ||
GenBank mRNA | AY094394.1 | - | 3 - 4 | 660 | 76 | |
BT001118.1 | - | 3 - 4 | 478 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |