Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335311.1 | NP_001325220.1 | 6 - 7 | 1390 | 112 | |
NM_126681.3 | NP_178722.1 | 6 - 7 | 1192 | 112 | ||
GenBank mRNA | AY094394.1 | - | 6 - 7 | 1105 | 112 | |
BT001118.1 | - | 6 - 7 | 923 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |