Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 19 - 20 | 2067 | 93 | |
NM_001335346.1 | NP_001323940.1 | 19 - 20 | 2067 | 93 | ||
NM_001335347.1 | NP_001323936.1 | 18 - 19 | 1984 | 93 | ||
NM_001335348.1 | NP_001323937.1 | 18 - 19 | 1913 | 93 | ||
NM_001335349.1 | NP_001323938.1 | 16 - 17 | 1844 | 93 | ||
NM_126770.8 | NP_178811.7 | 18 - 19 | 1924 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |