Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126855.4 | NP_565351.1 | 7 - 8 | 1672 | 92 | |
GenBank mRNA | AF367339.1 | - | 7 - 8 | 1557 | 92 | |
AY093990.1 | - | 6 - 7 | 1329 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |