Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335389.1 | NP_001325087.1 | 17 - 18 | 3454 | 94 | |
NM_001335390.1 | NP_001325086.1 | 17 - 18 | 2850 | 94 | ||
NM_126926.5 | NP_178970.3 | 17 - 18 | 3048 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |