Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335389.1 | NP_001325087.1 | 26 - 27 | 4839 | 117 | |
NM_001335390.1 | NP_001325086.1 | 26 - 27 | 4235 | 117 | ||
NM_126926.5 | NP_178970.3 | 26 - 27 | 4433 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |