Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GONST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202595.1 | NP_001189524.1 | 3 - 4 | 479 | 83 | |
NM_001335395.1 | NP_001318219.1 | 2 - 3 | 612 | 83 | ||
NM_126944.5 | NP_565357.1 | 2 - 3 | 603 | 83 | ||
NM_179621.3 | NP_849952.1 | 2 - 3 | 603 | 83 | ||
GenBank mRNA | BT030001.1 | - | 1 - 2 | 348 | 83 | |
KR265320.1 | - | 1 - 2 | 348 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |