Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GONST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202595.1 | NP_001189524.1 | 4 - 5 | 605 | 98 | |
NM_001335395.1 | NP_001318219.1 | 3 - 4 | 738 | 98 | ||
NM_001335396.1 | NP_001324974.1 | 2 - 3 | 1056 | 98 | ||
NM_001335397.1 | NP_001324975.1 | 1 - 2 | 398 | 98 | ||
NM_001335398.1 | NP_001324973.1 | 3 - 4 | 664 | 98 | ||
NM_126944.5 | NP_565357.1 | 3 - 4 | 729 | 98 | ||
NM_179621.3 | NP_849952.1 | 3 - 4 | 729 | 98 | ||
GenBank mRNA | BT030001.1 | - | 2 - 3 | 474 | 98 | |
KR265320.1 | - | 2 - 3 | 474 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |