Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GONST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202595.1 | NP_001189524.1 | 5 - 6 | 707 | 71 | |
NM_001335395.1 | NP_001318219.1 | 4 - 5 | 840 | 71 | ||
NM_001335396.1 | NP_001324974.1 | 3 - 4 | 1158 | 71 | ||
NM_001335397.1 | NP_001324975.1 | 2 - 3 | 500 | 71 | ||
NM_001335398.1 | NP_001324973.1 | 4 - 5 | 766 | 71 | ||
NM_126944.5 | NP_565357.1 | 4 - 5 | 831 | 71 | ||
NM_179621.3 | NP_849952.1 | 4 - 5 | 831 | 71 | ||
GenBank mRNA | BT030001.1 | - | 3 - 4 | 576 | 71 | |
KR265320.1 | - | 3 - 4 | 576 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |